Learn More
Invitrogen™ NHEDC2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5114007
Description
Immunogen sequence: IFISFAWLPK ATVQAAIGSV ALDTARSHGE KQLEDYGMDV LTVAFLSILI For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 93%; Dog: 86%; Guinea Pig: 83%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 86%; Rabbit: 93%; Rat: 100%.
Electroneutral exchange of protons for Na(+) and Li(+) across the inner mitochondrial membrane. Contributes to the organellar volume homeostasis. Required for osteoclast differentiation and bone resorption activity.
Specifications
| NHEDC2 | |
| Polyclonal | |
| Unconjugated | |
| SLC9B2 | |
| C80638; mitochondrial Na(+)/H(+) exchanger NHA2; mitochondrial sodium/hydrogen exchanger 9B2; mitochondrial sodium/hydrogen exchanger NHA2; Na(+)/H(+) exchanger NHA2; Na(+)/H(+) exchanger-like domain-containing protein 2; Na+/H+ exchanger domain containing 2; Nha2; nhaoc; nha-oc; NHE domain-containing protein 2; NHE10; Nhedc2; SLC9B2; Soditlum/hydrogen exchanger-like domain-containing protein 2; Sodium/hydrogen exchanger 9B2; Sodium/hydrogen exchanger-like domain-containing protein 2; solute carrier family 9 member B2; solute carrier family 9 subfamily B member 2; solute carrier family 9, subfamily B (cation proton antiporter 2), member 2; solute carrier family 9, subfamily B (NHA2, cation proton antiporter 2), member 2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 133308 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q86UD5 | |
| SLC9B2 | |
| Synthetic peptide directed towards the C terminal region of human NHEDC2 (aa 451-500). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.