Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NHEJ1 Monoclonal Antibody (OTI1F3), TrueMAB™, OriGene
SDP

Catalog No. 502541865
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-1865 30 μL
50-167-3095 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-1865 Supplier OriGene Supplier No. TA502003S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NHEJ1 Monoclonal antibody specifically detects NHEJ1 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen NHEJ1
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI1F3
Concentration 0.77 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene NHEJ1
Gene Accession No. Q9H9Q4
Gene Alias 1700029B21Rik; cernunnos; FLJ12610; Nhej1; non-homologous end joining factor 1; non-homologous end-joining factor 1; nonhomologous end-joining factor 1; protein cernunnos; Xlf; XRCC4-like factor
Host Species Mouse
Immunogen Full length human recombinant protein of human NHEJ1 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79840
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.