Learn More
NHEJ1 Mouse anti-Human, Clone: OTI3E6, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF502004

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Double-strand breaks in DNA result from genotoxic stresses and are among the most damaging of DNA lesions. This gene encodes a DNA repair factor essential for the nonhomologous end-joining pathway, which preferentially mediates repair of double-stranded breaks. Mutations in this gene cause different kinds of severe combined immunodeficiency disorders.Specifications
NHEJ1 | |
Monoclonal | |
Unconjugated | |
NHEJ1 | |
XLF | |
Mouse | |
Affinity Chromatography | |
RUO | |
79840 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
OTI3E6 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q9H9Q4 | |
NHEJ1 | |
Full length human recombit protein of human NHEJ1 produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.