Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NHERF-2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP18494425UL
Description
NHERF-2 Polyclonal specifically detects NHERF-2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
NHERF-2 | |
Polyclonal | |
Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
Q15599 | |
SLC9A3R2 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:RLVEVNGVNVEGETHHQVVQRIKAVEGQTRLLVVDQETDEELRRRQLTCTEEMAQRGLPPAHDPWEPKPDWAHTGSHSSEAGKKDVSGPLRELRPRLCHLRKGPQGYGFNLH | |
Affinity Purified | |
RUO | |
9351 | |
Human | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
E3KARP, Na(+)/H(+) exchange regulatory cofactor NHE-RF2, NHE3RF2, NHERF2MGC104639, NHERF-2NHE3 kinase A regulatory protein E3KARP, OCTS2, SIP-1SRY-interacting protein 1, sodium/hydrogen exchanger, Sodium-hydrogen exchanger regulatory factor 2, solute carrier family 9 (sodium/hydrogen exchanger), isoform 3 regulator 2, solute carrier family 9 (sodium/hydrogen exchanger), isoform 3 regulatoryfactor 2, solute carrier family 9 (sodium/hydrogen exchanger), member 3 regulator 2, Solute carrier family 9 isoform A3 regulatory factor 2, TKA-1SIP1, Tyrosine kinase activator protein 1 | |
Rabbit | |
37 kDa | |
25ul | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction