Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NHERF2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA550067
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA550067 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA550067 Supplier Invitrogen™ Supplier No. PA550067
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total SLC9A3R2 / NHERF2 protein.

NHE3 is a sodium/hydrogen exchanger in the brush border membrane of the proximal tubule, small intestine, and colon that plays a major role in transepithelial sodium absorption. SLC9A3R2, as well as SLC9A3R1 and protein kinase A phosphorylation, may play a role in NHE3 regulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NHERF2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene SLC9A3R2
Gene Accession No. Q15599, Q920G2, Q9JHL1
Gene Alias 0610011L07Rik; 1200011K07Rik; 2010007A20Rik; E3karp; HGNC:11076; MGC104639; Na(+)/H(+) exchange regulatory cofactor NHE-RF2; NHE3 kinase A regulatory protein E3KARP; NHE3 regulatory factor 2; NHE3RF2; Nherf2; NHERF-2; OCTS2; SIP1; SIP-1; SLC9A3 regulator 2; SLC9A3R2; sodium/hydrogen exchanger; sodium/hydrogen exchanger 3 kinase A regulatory protein; Sodium-hydrogen exchanger regulatory factor 2; solute carrier family 9 (sodium/hydrogen exchanger), isoform 3 regulator 2; solute carrier family 9 (sodium/hydrogen exchanger), isoform 3 regulatory factor 2; solute carrier family 9 (sodium/hydrogen exchanger), member 3 regulator 2; solute carrier family 9 isoform 3 regulator 2; solute carrier family 9 isoform 3 regulator 2, isoform A; sodium/hydrogen exchanger; solute carrier family 9 isoform A3 regulatory factor 2; solute carrier family 9, subfamily A (NHE3, cation proton antiporter 3), member 3 regulator 2; Sry interacting protein 1; SRY-interacting protein 1; Sryip1; TKA-1; tyrosine kinase activator protein 1
Gene Symbols SLC9A3R2
Host Species Rabbit
Immunogen Synthesized peptide derived from internal of human SLC9A3R2.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116501, 65962, 9351
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.