Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NHERF2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA581965
Description
Immunogen sequence: RLVEVNGVNV EGETHHQVVQ RIKAVEGQTR LLVVDQETDE ELRRRQLTCT EEMAQRGLPP AHDPWEPKPD WAHTGSHSSE AGKKDVSGPL RELRPRLCHL RKGPQGYGFN LH.
NHE3 is a sodium/hydrogen exchanger in the brush border membrane of the proximal tubule, small intestine, and colon that plays a major role in transepithelial sodium absorption. SLC9A3R2, as well as SLC9A3R1 and protein kinase A phosphorylation, may play a role in NHE3 regulation.
Specifications
NHERF2 | |
Polyclonal | |
Unconjugated | |
SLC9A3R2 | |
0610011L07Rik; 1200011K07Rik; 2010007A20Rik; E3karp; HGNC:11076; MGC104639; Na(+)/H(+) exchange regulatory cofactor NHE-RF2; NHE3 kinase A regulatory protein E3KARP; NHE3 regulatory factor 2; NHE3RF2; Nherf2; NHERF-2; OCTS2; SIP1; SIP-1; SLC9A3 regulator 2; SLC9A3R2; sodium/hydrogen exchanger; sodium/hydrogen exchanger 3 kinase A regulatory protein; Sodium-hydrogen exchanger regulatory factor 2; solute carrier family 9 (sodium/hydrogen exchanger), isoform 3 regulator 2; solute carrier family 9 (sodium/hydrogen exchanger), isoform 3 regulatory factor 2; solute carrier family 9 (sodium/hydrogen exchanger), member 3 regulator 2; solute carrier family 9 isoform 3 regulator 2; solute carrier family 9 isoform 3 regulator 2, isoform A; sodium/hydrogen exchanger; solute carrier family 9 isoform A3 regulatory factor 2; solute carrier family 9, subfamily A (NHE3, cation proton antiporter 3), member 3 regulator 2; Sry interacting protein 1; SRY-interacting protein 1; Sryip1; TKA-1; tyrosine kinase activator protein 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9351 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q15599 | |
SLC9A3R2 | |
Recombinant protein corresponding to Human SLC9A3R2. Recombinant protein control fragment (Product #RP-100916). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction