Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NHLH1 + NHLH2 Rabbit anti-Human∣Mouse∣Rat, Polyclonal, Bioss
SDP

Catalog No. 501981442
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1442 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1442 Supplier Bioss Supplier No. BS11523R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The helix-loop-helix (HLH) structures are known motifs commonly found in membrane-active and DNA-binding proteins. Helix-loop-helix proteins (HEN1 and HEN2) are DNA-binding proteins that may be involved in cell-type determination in the early nervous system. Studies of expression in normal tissues have demonstrated expression of NSCL-1 and NSCL-2 (genes encoding HEN1 and HEN2) in the developing central and peripheral nervous system, specifically in developing neurons.

Specifications

Antigen NHLH1 + NHLH2
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene NHLH1
Gene Accession No. Q02575, Q02576, Q02577, Q64221
Gene Alias 6230401I09Rik; BHLHA34; BHLHA35; Class A basic helix-loop-helix protein 34; Class A basic helix-loop-helix protein 35; Helix-loop-helix protein 1; HELIX-LOOP-HELIX PROTEIN 1 (HEN1) (NSCL); Helix-loop-helix protein 2; Hen1; HEN-1; Hen2; HEN-2; KIAA0490; Nescient helix loop helix 1; Nescient helix loop helix 2; nescient helix-loop-helix 1; nescient helix-loop-helix 2; NHLH1; Nhlh2; NSCL; NSCL1; NSCL-1; NSCL2; NSCL-2; Tal2
Gene Symbols NHLH1, NHLH2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human NHLH1 + NHLH2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18071, 18072, 289230, 295327, 4807, 4808
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.