Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NHLH2 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981443
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1443 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1443 Supplier Bioss Supplier No. BS11524R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

May serve as DNA-binding protein and may be involved in the control of cell-type determination, possibly within the developing nervous system.

Specifications

Antigen NHLH2
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene NHLH2
Gene Accession No. Q02577
Gene Alias 6230401I09Rik; BHLHA34; Class A basic helix-loop-helix protein 34; Helix-loop-helix protein 2; HEN2; HEN-2; KIAA0490; Nescient helix loop helix 2; nescient helix-loop-helix 2; NHLH2; NSCL2; NSCL-2
Gene Symbols NHLH2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human NHLH2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4808
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.