Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NHLRC1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA576985
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
Progressive myoclonic epilepsy type 2 (EPM2), also called Lafora disease, is an autosomal recessive disease characterized by grand mal seizures and/or myoclonus at about 15 years of age. Rapid and severe mental deterioration follows, often with psychotic features. Survival is less than 10 years after onset. Starch-like, endoplasmic reticulum-associated polyglucosans, called Lafora bodies, can be observed in brain, muscle, liver and heart. One cause of Lafora disease is due to mutations in NHLRC1, the gene encoding Malin. Forty-nine different mutations in NHLRC1 have been shown to cause EPM2. Malin, also called NHL repeat-containing protein 1, is a single subunit E3 ubiquitin ligase, containing 6 NHL repeats and 1 RING-type zinc finger. Malin's RING domain is responsible for its ability to mediate ubiquitination. Malin interacts with and polyubiquitinates Laforin, a protein also implicated in EPM2. Malin localizes to the endoplasmic reticulum and, to a lesser extent, in the nucleus. Malin is expressed in brain, cerebellum, spinal cord, medulla, heart, liver, skeletal muscle and pancreas.
Specifications
NHLRC1 | |
Polyclonal | |
Unconjugated | |
NHLRC1 | |
AI505271; B230309E09Rik; bA204B7.2; E3 ubiquitin-protein ligase NHLRC1; EPM2A; EPM2B; Malin; NHL repeat containing 1; NHL repeat containing E3 ubiquitin protein ligase 1; NHL repeat-containing protein 1; NHLRC1; RING-type E3 ubiquitin transferase NHLRC1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
105193, 364682, 378884 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.2 | |
Q6IMG5, Q6VVB1, Q8BR37 | |
NHLRC1 | |
Recombinant full length Human NHLRC1. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction