Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NHLRC2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566090
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566090 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566090 Supplier Invitrogen™ Supplier No. PA566090
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TGHHRILVVWK NGQIQYSIGG PNPGRKDGIF SESTFNSPQG VAIMNNIIYV ADTENHLIRK IDLEAEKVST VAGIGIQGTD KEGGAKGEQQ PIS Highest antigen sequence identity to the following orthologs - mouse 83%, rat 85%.

NHLRC2 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NHLRC2
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NHLRC2
Gene Accession No. Q8NBF2
Gene Alias 1200003G01Rik; AI835049; AV002846; AW496455; NHL repeat containing 2; NHL repeat-containing protein 2; Nhlrc2; novel NHL repeat domain containing protein
Gene Symbols NHLRC2
Host Species Rabbit
Immunogen Recombinant Human NHLRC2. Recombinant protein control fragment (Product #RP-106787).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 374354
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.