Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NHLRC3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA569332
Description
This target displays homology in the following species: Cow: 86%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 93%; Rabbit: 100%; Rat: 100%; Zebrafish: 75%.
This gene encodes a protein containing NCL-1, HT2A and Lin-41 (NHL) family repeats. Mammalian NHL-repeat containing proteins may be involved in a variety of enzymatic processes, including protein modification through ubiquitination. Alternatively spliced transcript variants have been observed for this gene.
Specifications
NHLRC3 | |
Polyclonal | |
Unconjugated | |
NHLRC3 | |
4833441N19; 8030451K01Rik; mKIAA4083; NHL repeat containing 3; NHL repeat-containing protein 3; NHLRC3; RGD1565414 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
387921 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q5JS37 | |
NHLRC3 | |
synthetic peptide directed towards the middle region of human NHLRC3. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction