Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NHLRC3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553082
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553082 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553082 Supplier Invitrogen™ Supplier No. PA553082
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ILWLHGENGT GPAKFNIPHS VTLDSAGRVW VADRGNKRIQ VFDKDTGEWL GAWNNCFTEE GPSSVRFTPD GKYLIVAQLN LSRLSVVAAP PVGSIGECSV ISTIQLADQ Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 88%.

This gene encodes a protein containing NCL-1, HT2A and Lin-41 (NHL) family repeats. Mammalian NHL-repeat containing proteins may be involved in a variety of enzymatic processes, including protein modification through ubiquitination. Alternatively spliced transcript variants have been observed for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NHLRC3
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NHLRC3
Gene Accession No. Q5JS37
Gene Alias 4833441N19; 8030451K01Rik; mKIAA4083; NHL repeat containing 3; NHL repeat-containing protein 3; NHLRC3; RGD1565414
Gene Symbols NHLRC3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NHLRC3. Recombinant protein control fragment (Product #RP-90609).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 387921
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.