Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NHP2L1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | NHP2L1 |
---|---|
Dilution | Western Blot 0.4 ug/ml, Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 |
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
NHP2L1 Polyclonal specifically detects NHP2L1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
NHP2L1 | |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human | |
[U4/U6.U5] tri-snRNP 15.5 kD RNA binding protein, 15.5K, FA1, FA-1, High mobility group-like nuclear protein 2 homolog 1, hSNU13, NHP2 non-histone chromosome protein 2-like 1 (S. cerevisiae), NHP2-like protein 1, NHPX, non-histone chromosome protein 2 (S. cerevisiae)-like 1, non-histone chromosome protein 2-like 1, OTK27, small nuclear ribonucleoprotein 15.5kDa (U4/U6.U5), SNRNP15-5, SNU13, SNU13 homolog, SPAG12, sperm specific antigen 1, SSFA1, U4/U6.U5 tri-snRNP 15.5 kDa protein | |
SNU13 | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Western Blot 0.4 ug/ml, Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
Polyclonal | |
Rabbit | |
DNA Repair, DNA replication Transcription Translation and Splicing | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
4809 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:TEADVNPKAYPLADAHLTKKLLDLVQQSCNYKQLRKGANEATKTLNRGISEFIVMAADAEPLEIILHLPLLCEDKNVPYVFVRSKQALGRACGVSRPVIACSVTIKEGSQLKQQIQSIQQSIERL | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title