Learn More
Invitrogen™ NHP2L1 Monoclonal Antibody (5C5)

Description
Peptide Sequence: MTEADVNPKA YPLADAHLTK KLLDLVQQSC NYKQLRKGAN EATKTLNRGI SEFIVMAADA EPLEIILHLP LLCEDKNVPY VFVRSKQALG RACGVSRPVI.
Specifications
Specifications
| Antigen | NHP2L1 |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 5C5 |
| Concentration | 0.22 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | SNU13 |
| Gene Accession No. | P55769 |
| Gene Alias | [U4/U6.U5] tri-snRNP 15.5 kD RNA binding protein; 15.5K; CTA-216E10.8; FA1; FA-1; Fertilization antigen 1; fertilization antigen-1; Fta1; high mobility group-like nuclear protein 2 homolog 1; hSNU13; NHP2 non-histone chromosome protein 2-like 1; NHP2 non-histone chromosome protein 2-like 1 (S. cerevisiae); NHP2L1; NHP2-like protein 1; NHP2-like protein 1, N-terminally processed; NHPX; non-histone chromosome protein 2-like 1; Otk27; small nuclear ribonucleoprotein 15.5kDa (U4/U6.U5); SNRNP15-5; SNU13; SNU13 homolog; SNU13 homolog, small nuclear ribonucleoprotein (U4/U6.U5); SPAG12; sperm specific antigen 1; sperm-specific antigen 1; Ssfa1; U4/U6.U5 small nuclear ribonucleoprotein SNU13; U4/U6.U5 tri-snRNP 15.5 kDa protein; U4/U6.U5] tri-snRNP 15.5 kDa protein |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.