Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NHP2L1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA522010
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA522010 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA522010 Supplier Invitrogen™ Supplier No. PA522010
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, H1299, HeLa, HepG2, Molt-4, Raji. Predicted reactivity: Xenopus laevis (92%), Pig (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Originally named because of its sequence similarity to the Saccharomyces cerevisiae NHP2 (non-histone protein 2), this protein appears to be a highly conserved nuclear protein that is a component of the [U4/U6.U5] tri-snRNP. It binds to the 5' stem-loop of U4 snRNA. Two transcript variants encoding the same protein have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NHP2L1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene SNU13
Gene Accession No. P55769, Q9D0T1
Gene Alias [U4/U6.U5] tri-snRNP 15.5 kD RNA binding protein; 15.5K; CTA-216E10.8; FA1; FA-1; Fertilization antigen 1; fertilization antigen-1; Fta1; high mobility group-like nuclear protein 2 homolog 1; hSNU13; NHP2 non-histone chromosome protein 2-like 1; NHP2 non-histone chromosome protein 2-like 1 (S. cerevisiae); NHP2L1; NHP2-like protein 1; NHP2-like protein 1, N-terminally processed; NHPX; non-histone chromosome protein 2-like 1; Otk27; small nuclear ribonucleoprotein 15.5kDa (U4/U6.U5); SNRNP15-5; SNU13; SNU13 homolog; SNU13 homolog, small nuclear ribonucleoprotein (U4/U6.U5); SPAG12; sperm specific antigen 1; sperm-specific antigen 1; Ssfa1; U4/U6.U5 small nuclear ribonucleoprotein SNU13; U4/U6.U5 tri-snRNP 15.5 kDa protein; U4/U6.U5] tri-snRNP 15.5 kDa protein
Gene Symbols Nhp2l1, SNU13
Host Species Rabbit
Immunogen Full length human NHP2L1 Recombinant protein.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20826, 300092, 4809
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.