Learn More
Invitrogen™ NHP2L1 Polyclonal Antibody

Description
Immunogen sequence: TEADVNPKAY PLADAHLTKK LLDLVQQSCN YKQLRKGANE ATKTLNRGIS EFIVMAADAE PLEIILHLPL LCEDKNVPYV FVRSKQALGR ACGVSRPVIA CSVTIKEGSQ LKQQIQSIQQ SIERL Highest antigen sequence indentity to the following orthologs: Rat - 100%, Mouse - 100%.
Specifications
Specifications
| Antigen | NHP2L1 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | SNU13 |
| Gene Accession No. | P55769 |
| Gene Alias | [U4/U6.U5] tri-snRNP 15.5 kD RNA binding protein; 15.5K; CTA-216E10.8; FA1; FA-1; Fertilization antigen 1; fertilization antigen-1; Fta1; high mobility group-like nuclear protein 2 homolog 1; hSNU13; NHP2 non-histone chromosome protein 2-like 1; NHP2 non-histone chromosome protein 2-like 1 (S. cerevisiae); NHP2L1; NHP2-like protein 1; NHP2-like protein 1, N-terminally processed; NHPX; non-histone chromosome protein 2-like 1; Otk27; small nuclear ribonucleoprotein 15.5kDa (U4/U6.U5); SNRNP15-5; SNU13; SNU13 homolog; SNU13 homolog, small nuclear ribonucleoprotein (U4/U6.U5); SPAG12; sperm specific antigen 1; sperm-specific antigen 1; Ssfa1; U4/U6.U5 small nuclear ribonucleoprotein SNU13; U4/U6.U5 tri-snRNP 15.5 kDa protein; U4/U6.U5] tri-snRNP 15.5 kDa protein |
| Gene Symbols | SNU13 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.