Learn More
Description
This product should be dissolved in DMSO (solubility = 100 mM) or DMF prior to further dilution in aqueous buffers.
Specifications
Specifications
| Chemical Name or Material | 6-[[5-[(3 AS,4 S,6 aR)-Hexahydro-2-oxo-1 H-thieno[3,4-d]imidazol-4-yl]-1-oxopentyl]amino]hexanoic acid 2,5-dioxo-1-pyrrolidinyl ester |
| CAS | 72040-63-2 |
| Quantity | 50 mg |
| Product Line | Tocris |
| Target | Buffers, Chelators and Reagents |
| Molecular Formula | C20H30N4O6S |
| Purity | 0.9 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
