Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NHS Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556760
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556760 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556760 Supplier Invitrogen™ Supplier No. PA556760
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TDLPYLEEST LTTAALSPSK IRPHTANKSV SRQYSTEDTI LSFLDSSAVE MGPDKLHLEK NSTFDVKNRC DPETITSAGS SLLDSNVTKD QVRTETEPIP ENTPTKNCAF PTEGFQRVSA ARPNDLDGKI IQYGPGPDET LEQ Highest antigen sequence identity to the following orthologs: Mouse - 64%, Rat - 63%.

May function in cell morphology by maintaining the integrity of the circumferential actin ring and controlling lamellipod formation. Involved in the regulation eye, tooth, brain and craniofacial development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NHS
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NHS
Gene Accession No. Q6T4R5
Gene Alias congenital cataracts and dental anomalies protein; CTRCT40; CXN; Nance-Horan syndrome (congenital cataracts and dental anomalies); Nance-Horan syndrome protein; NHS; NHS actin remodeling regulator; SCML1
Gene Symbols NHS
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NHS. Recombinant protein control fragment (Product #RP-92106).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4810
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.