Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Ni Sepharose™ 6 Fast Flow
GSA_VA

Catalog No. 45002986 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
100 mL
500 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-002-986 500 mL
45-002-987 5 mL
45-002-984 25 mL
45-002-985 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 45-002-986 Supplier Cytiva Supplier No. 17531803
Add to Cart
Add to Cart
Request a Quote

Combines the advantages of Ni2 for purification of histidine tagged proteins, with the well industrially established properties of the Sepharose Fast Flow platform. Cytiva Lifescience™ Ni Sepharose™ 6 Fast Flow is part of a range of IMAC chromatography media for capture and intermediate purification.

Ni Sepharose 6 Fast Flow is a member of the Cytiva range of IMAC chromatography media for capture and intermediate purification.

  • Designed for scaling up purification of histidine-tagged proteins.
  • High protein binding capacity and minimal leakage of Ni2+ ions.
  • As a BioProcess medium it fulfills industrial demands for security of supply, robust performance and regulatory support.
  • Available in convenient prepacked HisPrep and HisTrap column formats.
  • Composed of cross-linked 6% agarose beads modified with a novel chelating ligand immobilized to the base matrix and pre-charged with Ni2+ ions for purification of histidine tagged proteins.
  • The coupling chemistry and the novel ligand is designed to give minimal ligand and Ni2+ leakage while providing high capacity.
  • Available in a range of different bulk pack sizes and convenient pre-packed formats for easy scale-up and process development.
  • As member of the BioProcess media range, Ni Sepharose 6 Fast Flow meets industrial demands with security of supply and comprehensive technical and regulatory support.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Buffer 0.1M HCl, 0.1M NaOH, 8M Urea
Product Type Ni Sepharose 6 FF chromatography media
Quantity 500 mL
Height (English) 9.84 in.
Height (Metric) 250mm
Flow Rate 600 cm/hr.
Max. Pressure 1 bar
pH Range 3 to 12
Particle Size 90μm
Support Type 6% Cross-linked Agarose
Content And Storage 4° to 30°C
Matrix Ni-Sepharose
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.