Learn More
Invitrogen™ Niban-like protein Polyclonal Antibody

Description
Immunogen sequence: EVTDMNLNVI NEGGIDKLGE YMEKLSRLAY HPLKMQSCYE KMESLRLDGL QQRFDVSSTS VFKQRAQIHM REQMDNAVYT FETLLHQELG KGPTKEELCK SIQRV Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%.
Specifications
Specifications
| Antigen | Niban-like protein |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | Fam129b |
| Gene Accession No. | Q96TA1 |
| Gene Alias | 9130404D14Rik; bA356B19.6; C9orf88; FAM129B; family with sequence similarity 129 member B; family with sequence similarity 129, member B; Meg-3; Melanoma invasion by ERK; MINERVA; NIBAN2; Niban-like protein 1; OC58; Protein FAM129B; Protein Niban 2; RGD1307018 |
| Gene Symbols | Fam129b |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.