Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Niban-like protein Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554403
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554403 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554403 Supplier Invitrogen™ Supplier No. PA554403
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LIGNSLPGTT AKSGSAPILK CPTQFPLILW HPYARHYYFC MMTEAEQDKW QAVLQDCIRH CNNGIPEDSK VEGPAFTDAI RMYRQSKELY GTWEMLCGNE Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 94%.

FAM129B (Family With Sequence Similarity 129 Member B, Niban Like Protein 1) is a Protein Coding gene. May play a role in apoptosis suppression. May promote melanoma cell invasion in vitro. Among its related pathways are MAP Kinase Signaling. An important paralog of this gene is FAM129A.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Niban-like protein
Applications Western Blot, Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Fam129b
Gene Accession No. B4F7E8, Q8R1F1, Q96TA1
Gene Alias 9130404D14Rik; bA356B19.6; C9orf88; FAM129B; family with sequence similarity 129 member B; family with sequence similarity 129, member B; Meg-3; Melanoma invasion by ERK; MINERVA; NIBAN2; Niban-like protein 1; OC58; Protein FAM129B; Protein Niban 2; RGD1307018
Gene Symbols Fam129b
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human Niban-like protein. Recombinant protein control fragment (Product #RP-92964).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 227737, 362115, 64855
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.