Learn More
NIBL Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A301338A
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse and Rat
The Niban-like protein is encoded by a maternal imprinting gene. This protein may possess growth suppressor activity. Two major alternatively spliced isoforms have been described. forms. One form, Meg3-proximal (Meg3p), contains exons 1-3. The second form, Meg3-distal (Meg3d) did not contain exons 1-3 and was present in oocytes and in 1- and 2-cell embryos. Adachi H, Majima S, et al. Oncogene. 2004 Apr 22;23(19):3495-500 Matsumoto F, Fujii H, et al. Hum Pathol. 2006 Dec; 37(12):1592-600.Specifications
| NIBL | |
| Polyclonal | |
| Unconjugated | |
| FAM129B | |
| C9orf88 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 64855 | |
| 4° C | |
| Liquid |
| Western Blot | |
| 0.2 mg/ml | |
| TBS with 0.1% BSA and 0.09% sodium azide | |
| Q96TA1 | |
| FAM129B | |
| Between 683 and 733 | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.