Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NIBP Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA523130
Description
NIBP Polyclonal Antibody for Western Blot, ICC/IF, IP
NIBP is a 1246 amino acid intracellular regulatory protein. Reports suggest that NIBP physically interacts with NIK, IKK (beta), but not IKK (alpha) or IKK (gamma) and thus acts as a novel enhancer of cytokine-induced NF-(kappa) B signaling pathway. Due to its neuronal expression NIBP is considered as a potential target for modulating the NF-(kappa) B signaling cascade in neuronal pathologies dependent upon abnormal activation of this pathway. NIBP is mainly expressed in brain, muscle, heart, and kidney.
Specifications
NIBP | |
Polyclonal | |
Unconjugated | |
TRAPPC9 | |
1810044A24Rik; 2900005P22Rik; 4632408O18Rik; hypothetical protein LOC533451; Ibp; IKBKBBP; IKK2 binding protein; Kiaa1882; mKIAA1882; MRT13; Nibp; NIK- and IKBKB-binding protein; NIK and IKK(beta) binding protein; NIK and IKK{beta} binding protein; NIK and IKK-beta binding protein; RGD1309461; T1; trafficking protein particle complex 9; trafficking protein particle complex subunit 9; TRAPP 120 kDa subunit; TRAPPC9; TRS120; TRS130; tularik gene 1 protein | |
Rabbit | |
100 μL | |
Primary | |
Human, Mouse, Canine, Monkey, Bovine, Chicken | |
Antibody | |
IgG |
Immunoprecipitation, Western Blot, Immunocytochemistry | |
Conc. Not Determined | |
Whole serum with 50% glycerol and 0.025% sodium azide | |
Q32PH0, Q3U0M1, Q96Q05 | |
TRAPPC9 | |
Synthetic peptide (282 KKDFVGLDTDSRHYKKRC 299) of NIBP protein. | |
RUO | |
420312, 482058, 533451, 76510, 83696 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction