Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nicalin Antibody, DyLight 650, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP318435C
Description
Nicalin Polyclonal antibody specifically detects Nicalin in Human, Mouse samples. It is validated for Western BlotSpecifications
Nicalin | |
Polyclonal | |
Western Blot | |
NET59, nicalin, nicalin homolog (zebrafish), nicalin homolog (zebrefish), Nicastrin-like protein | |
Recognizes region between residues Between Between 375 and 425.. | |
0.1 mL | |
Neuroscience | |
56926 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
DyLight 650 | |
50mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction