Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nicalin/NCLN Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981500
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1500 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1500 Supplier Bioss Supplier No. BS11594R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Cyclin-dependent kinase 6 (p40cdk6 also known as PLSTIRE) belongs to a family of proteins designated as cdks. They are critical regulators of cell cycle progression. The prototype member of this family, p34cdc2, and a related protein cdk2, function late in the cycle while cdk4 and cdk6 are critically involved in G1 to S progression. CDK6 is essential for cell proliferation within the dentate gyrus of the hippocampus and the cuventricular zone of the lateral ventricles. Mutations in the gene results in microcephaly 12, primary, autosomal recessive (MCPH12).

Specifications

Antigen Nicalin/NCLN
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Ncln
Gene Accession No. Q969V3
Gene Alias 3100002P13Rik; AW120537; NCLN; NET59; Nicalin; nicalin homolog; nicalin homolog (zebrafish); Nicastrin-like protein; RGD1309355
Gene Symbols Ncln
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Nicalin.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56926
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.