Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Nicarbazin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA175315
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA175315 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA175315 Supplier Invitrogen™ Supplier No. PA175315
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

Working Concentration: 0.078 μg/mL. Sensitivity: 10 ng/mL Nicarbazin produces 55% inhibition in a competitive ELISA employing Nicarbazin polyclonal antibody.

Nicarbazin is an equimolar complex of 4,4'-Dinitrocarbanilide and 2-Hydroxy-4,6-dimethylpyrimidine. It is used as a coccidiostat for poultry. [National Center for Biotechnology Information. PubChem Compound Database; CID=9507].
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nicarbazin
Applications ELISA
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.1
Gene Alias C19H18N6O6; Nicarbazine; Nicrazin; Nicrazine
Host Species Sheep
Immunogen Nitrobenzene(4)-BTG.
Purification Method purified
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Target Molecule Chemical
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.