Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nicastrin Antibody (004), Alexa Fluor™ 488, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289826AF488
Description
Nicastrin Monoclonal specifically detects Nicastrin in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| Nicastrin | |
| Monoclonal | |
| Alexa Fluor 488 | |
| anterior pharynx-defective 2, ATAG1874, KIAA0253APH2, nicastrin | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human Nicastrin (NP_056146.1; Met1-Glu669). | |
| 0.1 mL | |
| Neurodegeneration, Neuronal Cell Markers, Neuroscience, Signal Transduction | |
| 23385 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunofluorescence | |
| 4 | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction