Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nicastrin Antibody (019), Alexa Fluor™ 700, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289828AF700
Description
Nicastrin Monoclonal specifically detects Nicastrin in Human samples. It is validated for Western Blot.Specifications
Nicastrin | |
Monoclonal | |
Alexa Fluor 700 | |
anterior pharynx-defective 2, ATAG1874, KIAA0253APH2, nicastrin | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human Nicastrin (NP_056146.1; Met1-Glu669). | |
0.1 mL | |
Neurodegeneration, Neuronal Cell Markers, Neuroscience, Signal Transduction | |
23385 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
19 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction