Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Nicastrin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589872
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589872 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589872 Supplier Invitrogen™ Supplier No. PA589872
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RFLRARNISG VVLADHSGAF HNKYYQSIYD TAENINVSYP EWLSPEEDLN FVTDTAKALA DVATVLGRAL YELAGGTNFS DTVQADPQTV TRLLYGFLIK ANNSWFQSIL RQDLRSYLGD GPLQHYIAVS SPTNTTYVVQ YALANLTGTV VNLTREQCQD PSKVPSENKD LYEYSWVQGP LHSNETDRLP RCVRSTARLA RALSPAFELS QWSSTEYSTW TESRWKDIRA RIFLIASKEL E; Positive Samples: Mouse brain, Rat brain, Rat spleen; Cellular Location: Melanosome, Membrane, Single-pass type I membrane protein.

NCSTN (Nicastrin) is a gene that encodes a type I transmembrane glycoprotein that is an integral component of the multimeric gamma-secretase complex. The encoded protein cleaves integral membrane proteins, including Notch receptors and beta-amyloid precursor protein, and may be a stabilizing cofactor required for gamma-secretase complex assembly. The cleavage of beta-amyloid precursor protein yields amyloid beta peptide, the main component of the neuritic plaque and the hallmark lesion in the brains of patients with Alzheimer's disease; however, the nature of the encoded protein's role in Alzheimer's disease is not known for certain. Mutations in this gene are associated with familial acne inversa. A pseudogene of this gene is present on chromosome 21.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nicastrin
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 2.07 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene NCSTN
Gene Accession No. P57716, Q8CGU6, Q92542
Gene Alias 9430068N19Rik; AA727311; anterior pharynx-defective 2; Aph2; ATAG1874; D1Dau13e; Kiaa0253; mKIAA0253; NCSTN; Nct; Nicastrin; OTTHUMP00000031840; UNQ1874/PRO4317
Gene Symbols NCSTN
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 429-669 of human NCSTN (NP_0561461).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23385, 289231, 59287
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.