Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nicastrin Rabbit anti-Human, Clone: 4, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289826100UL
Description
Nicastrin Monoclonal specifically detects Nicastrin in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
Nicastrin | |
Monoclonal | |
Unconjugated | |
0.2 um filtered solution in PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Immunofluorescence | |
4 | |
Immunocytochemistry/Immunofluorescence 1:20-1:100 | |
anterior pharynx-defective 2, ATAG1874, KIAA0253APH2, nicastrin | |
Recombinant Human Nicastrin protein | |
100 μg | |
Neurodegeneration, Neuronal Cell Markers, Neuroscience, Signal Transduction | |
23385 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction