Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Niceritrol 98.0+%, TCI America™
SDP

Catalog No. N0974100MG
Click to view available options
Quantity:
25 mg
100 mg

Chemical Identifiers

CAS 5868-05-3
Molecular Formula C29H24N4O8
Molecular Weight (g/mol) 556.53
MDL Number MFCD00215982
InChI Key KUEUWHJGRZKESU-UHFFFAOYSA-N
Synonym Pentaerythritol Tetranicotinate
PubChem CID 4476
IUPAC Name [3-(pyridine-3-carbonyloxy)-2,2-bis(pyridine-3-carbonyloxymethyl)propyl] pyridine-3-carboxylate
SMILES C1=CC(=CN=C1)C(=O)OCC(COC(=O)C2=CN=CC=C2)(COC(=O)C3=CN=CC=C3)COC(=O)C4=CN=CC=C4

Specifications

Melting Point 165°C
Color White-Yellow
Quantity 100 mg
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Niceritrol
RTECSNumber QT1980000
TSCA No
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.