Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nickel, 99.97%, (trace metal basis), spheres, 5 to 11 mm, Thermo Scientific Chemicals

Catalog No. ac201672500
Click to view available options
Quantity:
250 g
Packaging:
Plastic bottle

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-02-0
Molecular Formula Ni
Molecular Weight (g/mol) 58.69
MDL Number MFCD00011137 MFCD06798735
InChI Key PXHVJJICTQNCMI-UHFFFAOYSA-N
Synonym raney alloy, fibrex, catalyst, particles, fibrex p, nickel, elemental, nichel italian, nickel, soluble salts, carbonyl powder, niccolum
PubChem CID 935
ChEBI CHEBI:28112
IUPAC Name nickel
SMILES [Ni]

Specifications

Melting Point 1455.0°C
Boiling Point 2732.0°C to 2837.0°C
CAS Min % 99.95
CAS Max % 100.0
Density 8.9
Quantity 250 g
Packaging Plastic bottle
Form Sphere
Assay Percent Range 99.97%
Fieser 12,335; 13,197; 17,296
Merck Index 15, 6583
Formula Weight 58.7
Percent Purity 99.97%
Grade Trace Metal Basis
Chemical Name or Material Nickel, p.a.
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement:
Suspected of causing cancer.
Causes damage to organs through prolonged or repeated exposure.
May cause an allergic skin reaction.
Precautionary Statement GHS P Statement:
Wear eye protection/face protection.
Use personal protective equipment as required.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.