Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nickel ammonium sulfate hexahydrate

Catalog No. p-4824827
Click to view available options
Quantity:
100 g
500 g

CAS: 7785-20-8 | Ni(NH4)2(SO4)2·6H2O

It is used in electroplating. Has limited use as dye mordants and is used in metal-finishing compositions. Nickel electrolyte for electroplating

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7785-20-8
Molecular Formula Ni(NH4)2(SO4)2·6H2O
MDL Number MFCD00150260
Synonym Ammonium nickel sulfate hexahydrate

Specifications

Color Green
Physical Form Large Crystals
Quantity 100 g
Solubility Information Soluble in ammonium sulphate. Insoluble in alcohol. Slightly soluble in water.
Formula Weight 394.99 (286.90 Anhydrous)
Odor Odorless
Appearance Green crystals
Density 1.923
Chemical Name or Material Nickel ammonium sulfate hexahydrate
Hazard Category H302+H332-H317-H334-H341-H350i-H360D-H372
Precautionary Statement P201-P202-P260-P264b-P270-P271-P272-P280g-P281-P285-P301+P312-P302+P352-P304+P340-P308+P313-P330-P333+P313-P342+P311-P363-P501c
DOTInformation Hazard Class: 9; Packaging Group: III
EINECSNumber 239-793-5
RTECSNumber WS6061000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.