Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nickel Carbonate, Basic, Tetrahydrate, Powder, 45%, Spectrum™ Chemical
SDP

Catalog No. 18611946 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
12 kg
Packaging:
Poly Bottle
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-611-946 125 g Poly Bottle
18-611-945 25 g Poly Bottle
18-611-947 500 g Poly Bottle
18-611-948 2.5 kg Poly Pail
18-611-949 12 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-611-946 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. N1045125GM
Add to Cart
Add to Cart

Chemical Identifiers

CAS 39430-27-8

Specifications

CAS Max % 1
Quantity 125 g
Percent Purity 45%
Grade Reagent
Packaging Poly Bottle

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.