Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nickel Chloride AR Crystal, Macron™

Catalog No. MK6376212 Shop All Avantor Products
Change view
Click to view available options
Quantity:
12 x 500 g
4 x 2.5 kg
Packaging:
POLYSTORMOR
Poly
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MK-6376-212 4 x 2.5 kg Poly
MK-6376-500 12 x 500 g POLYSTORMOR
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MK-6376-212 Supplier Avantor Supplier No. 637606

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Chemical Identifiers

CAS 7791-20-0
Molecular Formula NiCl2·6H2O

Specifications

Physical Form Crystalline
Quantity 4 x 2.5 kg
Formula Weight 237.7
Packaging Poly
Chemical Name or Material Nickel Chloride, 6-Hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.