Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nickel(II) 2-ethylhexanoate monoisopropoxide, 99+% (m.b.), 5% w/v in IPA, pkgd under Ar in rslble ChemSeal bottles, Thermo Scientific™

Catalog No. AA42380AC
Click to view available options
Quantity:
25 mL

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

  • Packaged under Argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD01863473
Synonym Nickel(II) 2-ethylhexano-isopropoxide

Specifications

Packaging ChemSeal™ bottles
Quantity 25 mL
UN Number UN1219
Sensitivity Moisture sensitive
Formula Weight 261
Percent Purity ≥99% (m.b.)
Concentration or Composition (by Analyte or Components) 5% w/v in isopropanol
Physical Form Liquid
Chemical Name or Material Nickel (II) 2-ethylhexanoate monoisopropoxide
Hazard Category H225-H317-H319-H336-H351-H361
Hazard Statement GHS H Statement
H225-H351-H361-H315-H319-H317-H336
Highly flammable liquid and vapor.
Suspected of causing cancer.
Suspected of damaging fertility or the unborn child.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause drowsiness or dizziness.
Precautionary Statement P201-P202-P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P272-P280-P281-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P333+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ISOPROPANOL
TSCA No
storageNote1
  • Packaged under Argon
Recommended Storage Ambient temperatures
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.