Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nickel(II) acetate hydrate, 99+%

Catalog No. p-4407667
Click to view available options
Quantity:
250 g
1 kg
5 kg

CAS: 6018-89-9 | C4H14NiO8 | 248.84 g/mol

Used in synthesis of polynuclear-nickel polyoxotungstate cluster compounds that are ideal for the design of molecular magnets .

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6018-89-9
Molecular Formula C4H14NiO8
Molecular Weight (g/mol) 248.84
MDL Number MFCD00066973
InChI Key OINIXPNQKAZCRL-UHFFFAOYSA-L
Synonym nickel ii acetate tetrahydrate, nickel acetate tetrahydrate, nickel diacetate tetrahydrate, diacetatonickel tetrahydrate, nickelous acetate tetrahydrate, unii-6soa8l0560, acetic acid, nickel +2 salt, tetrahydrate, nickel 2+ diacetate tetrahydrate, nickel acetate tetrahydrate nickel and nickel compounds, acetic acid, nickel 2+ salt, tetrahydrate
PubChem CID 62601
IUPAC Name nickel(2+) diacetate tetrahydrate
SMILES O.O.O.O.[Ni++].CC([O-])=O.CC([O-])=O

Specifications

Melting Point (decomposition)
Color White
Physical Form Crystalline
Quantity 250 g
Assay Percent Range 99+%
Solubility Information Soluble in water,alcohol
Formula Weight 176.79 (Anhydrous)
Odor Light
Appearance Green crystals
Chemical Name or Material Nickel(II) acetate hydrate
Hazard Category H302+H332-H317-H334-H335-H341-H350i-H360D-H372
Precautionary Statement P201-P202-P260-P264b-P270-P271-P272-P280g-P281-P285-P301+P312-P302+P352-P304+P340-P308+P313-P330-P333+P313-P342+P311-P363-P501c
DOTInformation DOT Class: 9, Packaging Group: III
EINECSNumber 206-761-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.