Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nickel(II) bromide, anhydrous, 99%

Catalog No. p-7080281
Click to view available options
Quantity:
50 g
250 g

CAS: 13462-88-9 | Br2Ni | 218.501 g/mol

Nickel(II) bromide is used as a source of the bromide ion. It is an important raw material and intermediate used in synthetic chemistry. Further, it is used as a catalyst in cross-coupling reactions.It is used in the preparation of paramagnetic trans-diaquabis(N,N-diethylethylenediamine) nickel(II) bromide and diamagnetic bis(N,N-diethylethylenediamine)nickel(II) bromide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13462-88-9
Molecular Formula Br2Ni
Molecular Weight (g/mol) 218.501
MDL Number MFCD00011141
InChI Key IPLJNQFXJUCRNH-UHFFFAOYSA-L
Synonym nickel bromide, nickel ii bromide, nickel dibromide, nickelous bromide, nickel bromide nibr2, nickel 2+ bromide, unii-41sgh8a2s6, nibr2, nickel ii bromide, anhydrous
PubChem CID 278492
IUPAC Name dibromonickel
SMILES [Ni](Br)Br

Specifications

Melting Point 963°C
Physical Form Crystalline
Quantity 50 g
Linear Formula NiBr2
UN Number UN3077
Merck Index 14,6502
Solubility Information Soluble in water,alcohol and ether.
Formula Weight 218.53
Percent Purity 99%
Odor Odorless
Sensitivity Hygroscopic
Density 5.098 g/mL at 27°C
Chemical Name or Material Nickel (II) bromide, Anhydrous
Show More Show Less
Hazard Category H317-H334-H335-H341-H350i-H360D-H372
Hazard Statement GHS H Statement
H334-H350-H360-H372-H341-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause cancer.
May damage fertility or the unborn child.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing genetic defects.
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P260-P264b-P270-P271-P272-P280g-P281-P285-P302+P352-P304+P340-P308+P313-P333+P313-P342+P311-P363-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 236-665-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.