Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nickel(II) carbonate hydroxide hydrate, for analysis, (ca 45-47% Ni)
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Green |
| Physical Form | Powder |
| Quantity | 250 g |
| Linear Formula | 2Ni(OH)2·xH2O |
| Merck Index | 13, 6527 |
| Solubility Information | Solubility in water: insoluble. Other solubilities: soluble in ammonia and in diluted acids |
| Formula Weight | 304.11 |
| Percent Purity | ≥45% (NI) |
| Grade | Analytical |
| Packaging | Plastic bottle |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Harmful if swallowed. Harmful if inhaled. May cause an allergic skin reaction. Very toxic to aquatic life with long lasting effects. May cause cancer by inhalation. Suspected of causing genetic defects |
| Precautionary Statement | GHS P Statement Wear eye protection/face protection. Use personal protective equipment as required. Avoid release to the environment. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. IF INHALED: Remove |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction