Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nickel(II) chloride, anhydrous, 99.99% (metals basis)

Catalog No. AA4344114
Click to view available options
Quantity:
5 g
25 g

CAS: 7718-54-9 | Cl2Ni | 129.593 g/mol

Nickel(II) chloride is used in electroplating, electroforming ,electroless plating, tea and coffee plantation, dye mordant. It is also used in semiconducting metal-containing polymer synthesis reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7718-54-9
Molecular Formula Cl2Ni
Molecular Weight (g/mol) 129.593
MDL Number MFCD00011142
InChI Key QMMRZOWCJAIUJA-UHFFFAOYSA-L
PubChem CID 24385
ChEBI CHEBI:34887
IUPAC Name dichloronickel
SMILES Cl[Ni]Cl

Specifications

Melting Point 1,001°C
Color Yellow
Physical Form Crystalline Powder
Quantity 25 g
Linear Formula NiCl2
UN Number UN3288
Merck Index 14,6505
Solubility Information It is soluble in water,ethylene glycol,ethanol, ammonium hydroxide. It is insoluble in ammonia, nitric acid.
Formula Weight 129.6
Percent Purity 99.99%
Odor Odorless
Sensitivity Hygroscopic
Density 3.55 g/mL
Assay (metals basis)
Chemical Name or Material Nickel (II) chloride, Anhydrous
Show More Show Less
Hazard Category H301+H331-H315-H317-H319-H334-H341-H350i-H360D-H372-H500
Hazard Statement GHS H Statement
H301-H331-H334-H350-H360-H372-H341-H315-H317
Toxic if swallowed.
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause cancer.
May damage fertility or the unborn child.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing genetic defects.
Causes skin irritation.
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P260-P264b-P270-P271-P272-P280-P281-P285-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P311-P330-P333+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.
EINECSNumber 231-743-0
RTECSNumber QR6475000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.