Learn More
Nickel(II) chloride, anhydrous, 99% (metals basis)
Description
Nickel(II) chloride is a mild Lewis acid used for the regioselective isomerization of dienols and selective reduction of alkenes to alkanes in presence of Lithium aluminum hydride. It is used as a precursor to nickel boride, finely divided nickel. It acts as a catalyst to prepare dialkyl arylphosphonates from phosphites and aryl iodide. It finds application in the electroplating of nickel onto other metal items. It is also used in the synthesis of semiconducting metal-containing polymers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 1001°C |
Color | Yellow to Orange |
Physical Form | Powder |
Quantity | 500 g |
Assay Percent Range | 99% (metals basis) |
Solubility Information | Soluble in water,ethylene glycol,ethanol and ammonium hydroxide. Insoluble in ammonia and nitric acid. |
Formula Weight | 129.6 |
Sensitivity | Hygroscopic |
Appearance | Yellow to orange powder |
Vapor Pressure | 1.33 hPa (1mm Hg) at 20°C |
Show More |
Safety and Handling
missing translation for 'healthHazard1' | H301+H331-H315-H317-H319-H334-H341-H350i-H360D-H372-H500 |
missing translation for 'healthHazard2' | Avoid exposure - obtain special instructions before use. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment. Refer to special instructions/Safety data sheets. |
missing translation for 'healthHazard3' | P201-P202-P260-P264b-P270-P271-P272-P280-P281-P285-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P311-P330-P333+P313-P362-P501c |
DOTInformation | Hazard Class: 6.1, Packing Group: III |
EINECSNumber | 231-743-0 |
RTECSNumber | QR6475000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.