Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nickel(II) chloride, anhydrous, 99% (metals basis)

Catalog No. p-4782027
Click to view available options
Quantity:
100 g
500 g

CAS: 7718-54-9 | Cl2Ni | 129.593 g/mol

Nickel(II) chloride is a mild Lewis acid used for the regioselective isomerization of dienols and selective reduction of alkenes to alkanes in presence of Lithium aluminum hydride. It is used as a precursor to nickel boride, finely divided nickel. It acts as a catalyst to prepare dialkyl arylphosphonates from phosphites and aryl iodide. It finds application in the electroplating of nickel onto other metal items. It is also used in the synthesis of semiconducting metal-containing polymers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7718-54-9
Molecular Formula Cl2Ni
Molecular Weight (g/mol) 129.593
MDL Number MFCD00011142
InChI Key QMMRZOWCJAIUJA-UHFFFAOYSA-L
PubChem CID 24385
ChEBI CHEBI:34887
IUPAC Name dichloronickel
SMILES Cl[Ni]Cl

Specifications

Melting Point 1001°C
Color Yellow to Orange
Physical Form Powder
Quantity 100 g
Assay Percent Range 99% (metals basis)
Solubility Information Soluble in water,ethylene glycol,ethanol and ammonium hydroxide. Insoluble in ammonia and nitric acid.
Formula Weight 129.6
Sensitivity Hygroscopic
Appearance Yellow to orange powder
Vapor Pressure 1.33 hPa (1mm Hg) at 20°C
Density 3.55 g/cm3
Chemical Name or Material Nickel(II) chloride, anhydrous
Show More Show Less
Hazard Category H301+H331-H315-H317-H319-H334-H341-H350i-H360D-H372-H500
Hazard Statement Avoid exposure - obtain special instructions before use. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment. Refer to special instructions/Safety data sheets.
Precautionary Statement P201-P202-P260-P264b-P270-P271-P272-P280-P281-P285-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P311-P330-P333+P313-P362-P501c
DOTInformation Hazard Class: 6.1, Packing Group: III
EINECSNumber 231-743-0
RTECSNumber QR6475000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.