Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Nickel (II) Chloride Hexahydrate
SCI_ED

Catalog No. S25442 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Reagent Grade
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25442 100 g Reagent Grade
S25442A 500 g Reagent Grade
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. S25442
Add to Cart
Add to Cart
This item is not returnable. View return policy

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 7791-20-0
Synonym Nickel Dichloride, Nickelous Chloride

Specifications

Color Green
Quantity 100 g
Assay Percent Range >95%
Linear Formula NiCl2·6H2O
Formula Weight 237.72
Grade Reagent
Packaging Reagent Grade
Density 3.55g/mL
Chemical Name or Material Nickel (II) Chloride, Hexahydrate
Safety handling Check Alert storage code Red
ShelfLife 1 year
DOTInformation DOT Class 6.1, III, : Poison
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.