Learn More
Nickel(II) chloride, ultra dry, 99.9% (metals basis)
Description
It is used in the synthesis of semiconducting metal-containing polymers in which the polypyrrole backbone has a conformational energy minimum and is nearly planar.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 1001°C |
Color | Yellow to Orange |
Physical Form | Powder |
Quantity | 5 g |
Assay Percent Range | 99.9% (metals basis) |
Solubility Information | Soluble in water. |
Formula Weight | 129.6 |
Odor | Odorless |
Sensitivity | Hygroscopic |
Appearance | Yellow to orange powder |
Show More |
Safety and Handling
Hazard Category | H301+H331-H315-H317-H319-H334-H341-H350i-H360D-H372-H500 |
Precautionary Statement | P201-P202-P260-P264b-P270-P271-P272-P280-P281-P285-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P311-P330-P333+P313-P362-P501c |
DOTInformation | DOT Class: 6.1; Packing Group: III |
EINECSNumber | 231-743-0 |
RTECSNumber | QR6475000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.