Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nickel(II) citrate hydrate, 98%

Catalog No. p-7050886
Click to view available options
Quantity:
100 g
500 g

Catalog No. AA3963422

Supplier: Thermo Scientific Chemicals 03963422

Only null left
Add to Cart

CAS: 6018-92-4 | C12H10Ni3O14 | 554.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6018-92-4
Molecular Formula C12H10Ni3O14
Molecular Weight (g/mol) 554.28
MDL Number MFCD00054366
InChI Key UPPLJLAHMKABPR-UHFFFAOYSA-H
Synonym nickel ii citrate hydrate
PubChem CID 131881812
IUPAC Name 2-hydroxypropane-1,2,3-tricarboxylic acid;nickel
SMILES [Ni++].[Ni++].[Ni++].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O.OC(CC([O-])=O)(CC([O-])=O)C([O-])=O

Specifications

Quantity 100 g
UN Number UN3077
Formula Weight 554.33 (Anhydrous)
Percent Purity ≥98%
Physical Form Chunks and/or Powder
Chemical Name or Material Nickel(II) citrate hydrate
Hazard Category H302-H317-H334-H341-H350-H360-H372
Hazard Statement GHS H Statement
H334-H350-H360-H372-H341-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause cancer.
May damage fertility or the unborn child.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing genetic defects.
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P260-P264b-P270-P272-P280g-P281-P285-P301+P312-P302+P352-P304+P341-P308+P313-P330-P333+P313-P342+P311-P363-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 227-873-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.