Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nickel(II) cyanide tetrahydrate

Catalog No. p-4824842
Click to view available options
Quantity:
10 g
50 g

For Metallurgy, Nickel-Plating | CAS: 13477-95-7 | C2H8N2NiO4 | 182.789 g/mol

Applied in metallurgy, nickel-plating industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13477-95-7
Molecular Formula C2H8N2NiO4
Molecular Weight (g/mol) 182.789
MDL Number MFCD00049490
InChI Key VHHVAACTKRUWEK-UHFFFAOYSA-N
Synonym nickel ii cyanide tetrahydrate, ni cn 2 tetrahydrate, acmc-20ak3a, nickel cyanide tetrahydrate, c2n2ni.4h2o, nickel dicyanide tetrahydrate
PubChem CID 71317429
IUPAC Name nickel(2+);dicyanide;tetrahydrate
SMILES [C-]#N.[C-]#N.O.O.O.O.[Ni+2]

Specifications

Melting Point 200°C-4H2O
Color Green
Physical Form Powder
Quantity 10 g
Solubility Information Insoluble in water. Soluble in ammonium carbonate,ammonia,alkali cyanides
Formula Weight 182.73 (110.73 Anhydrous)
Appearance Light green powder
Chemical Name or Material Nickel(II) cyanide tetrahydrate
Hazard Category H317-H334-H350-H372
Precautionary Statement P201-P202-P260-P264b-P270-P272-P280g-P281-P285-P302+P352-P304+P341-P308+P313-P333+P313-P342+P311-P363-P501c
DOTInformation DOT Class: 6.1, Packaging Group: II
EINECSNumber 209-160-8
RTECSNumber QR6495000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.