Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nickel(II) formate dihydrate

Catalog No. AA1380136
Click to view available options
Quantity:
100 g
500 g
2 kg

For Nickel Catalyst Preparations | CAS: 15694-70-9 | Ni(HCO2)2·2H2O

Nickel(II) formate dihydrate is used in nickel catalyst preparations. It is also used as a biochemical for proteomics research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15694-70-9
Molecular Formula Ni(HCO2)2·2H2O
MDL Number MFCD00153034

Specifications

Melting Point 130°C to 140°C-2H2O
Color Green
Physical Form Powder
Quantity 500 g
Solubility Information In the range 180-200° decomposes to Ni,CO,CO2,H2,H2O,CH4. Partially soluble in water
Formula Weight 184.77 (148.74anhydrous)
Appearance Green powder
Density 2.15
Chemical Name or Material Nickel(II) formate dihydrate
Hazard Category H317-H334-H341-H350-H360-H372
Hazard Statement Avoid exposure - obtain special instructions before use. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment. Refer to special instructions/Safety data sheets.
Precautionary Statement P201-P202-P260-P264b-P270-P272-P280g-P281-P285-P302+P352-P304+P341-P308+P313-P333+P313-P342+P311-P363-P501c
DOTInformation Hazard Class: 9, Packing Group: III
EINECSNumber 222-101-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.