Alfa Aesar™ Nickel(II) oxide, 99.995% (metals basis)

Vacuum deposition

Manufacturer: Alfa Aesar™ 4442322

Quantity 100g

 View more versions of this product

Catalog No. AA4442322

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1313-99-1
    Molecular Formula NiO
    Formula Weight 74.71
    MDL Number MFCD00011145
    Merck Index 14,6512
    Chemical Name or Material Nickel (II) oxide

    Specifications

    Formula Weight 74.71
    Health Hazard 3 GHS P Statement
    P260-P261-P280-P363-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Wash contaminated clothing before reuse.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Density 6.67g/mL
    Odor Odorless
    Molecular Formula NiO
    Merck Index 14,6512
    Quantity 100g
    Health Hazard 1 Danger
    TSCA TSCA
    Particle Size 3 to 12mm
    Physical Form Pieces
    Color Green to black
    Melting Point 1984°C
    MDL Number MFCD00011145
    Health Hazard 2 GHS H Statement
    H350-H372-H317
    May cause cancer.
    Causes damage to organs through prolonged or repeated exposure.
    May cause an allergic skin reaction.
    RTECS Number QR8400000
    CAS 1313-99-1
    Chemical Name or Material Nickel (II) oxide
    Assay 99.995%, (metals basis)

    GHS H Statement
    H350-H372-H317
    May cause cancer.
    Causes damage to organs through prolonged or repeated exposure.
    May cause an allergic skin reaction.

    GHS P Statement
    P260-P261-P280-P363-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Wash contaminated clothing before reuse.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit