Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nickel(II) oxide, black, Ni 76%, (contains Ni{2}O{3})

Catalog No. p-7080367
Click to view available options
Quantity:
250 g
1 kg

CAS: 1313-99-1 | NiO | 74.69 g/mol

Nickel(II) oxide is used in fuel cell electrodes, storage batteries, electronic circuit boards, electrical ceramics, and as a pigment for ceramics and glass.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1313-99-1
Molecular Formula NiO
Molecular Weight (g/mol) 74.69
MDL Number MFCD00011145
InChI Key GNRSAWUEBMWBQH-UHFFFAOYSA-N
Synonym nickel ii oxide, nickel monoxide, bunsenite, nickelous oxide, nickel oxide, mononickel oxide, nickel protoxide, black nickel oxide, nickel 2+ oxide
PubChem CID 14805
IUPAC Name oxonickel
SMILES O=[Ni]

Specifications

Melting Point 1984°C
Color Black
Physical Form Powder
Quantity 250 g
Merck Index 14,6512
Solubility Information Soluble in acids. Insoluble in water.
Formula Weight 74.71
Concentration or Composition (by Analyte or Components) Ni 76%, (contains Ni2O3)
Odor Odorless
Density 6.67 g/mL
Chemical Name or Material Nickel (II) oxide
Show More Show Less
Hazard Category H317-H350i-H372
Hazard Statement GHS H Statement
H350-H372-H317
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P260-P264b-P270-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c
EINECSNumber 215-215-7
RTECSNumber QR8400000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.