Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nickel(II) perchlorate hydrate, 99.998% (metals basis)

Catalog No. p-7059278
Click to view available options
Quantity:
10 g
50 g

CAS: 13637-71-3 | Cl2NiO8 | 257.59 g/mol

Applied as a starting material for homometallic, trinuclear heteroscorpionate complexes applied in studies of electronic and magnetic properties.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13637-71-3
Molecular Formula Cl2NiO8
Molecular Weight (g/mol) 257.59
MDL Number MFCD00149804
InChI Key ZLQBNKOPBDZKDP-UHFFFAOYSA-L
Synonym nickel perchlorate, unii-bq8a24mezo, perchloric acid, nickel 2+ salt, nickel ii perchlorate, nickel 2+ diperchlorate, bq8a24mezo, perchloric acid, nickel 2+ salt 2:1, nickelous perchlorate, perchloric acid, nickel salt, nickel perchlorate ni clo4 2
PubChem CID 26158
SMILES [Ni++].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O

Specifications

Melting Point 140°C
Color Green
Physical Form Crystalline
Quantity 10 g
Linear Formula Ni(ClO4)2·xH2O
UN Number UN1481
Solubility Information Very soluble in water. Soluble in many organic solvents.
IUPAC Name nickel(2+) diperchlorate
Formula Weight 257.59 (Anhydrous)
Percent Purity 99.998%
Odor Odorless
Refractive Index 1.55
Sensitivity Hygroscopic
Assay (metals basis)
Chemical Name or Material Nickel (II) perchlorate hydrate
Show More Show Less
Hazard Category H272-H314-H317-H334-H335-H341-H350-H360-H372
Hazard Statement GHS H Statement
H272-H334-H350-H360-H372-H341-H314-H318-H317
May intensify fire
oxidizer.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause cancer.
May damage fertility or the unborn child.
Precautionary Statement P201-P202-P210-P220-P221-P260-P264b-P270-P271-P272-P280g-P281-P285-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P333+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: PERCHLORATES, INORGANIC, N.O.S.
EINECSNumber 237-124-1
RTECSNumber SC9550000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.